Order Entry
Slovenia
ContactUsLinkComponent
InfinityLab Quick Change valve head, 6-column selector, maximum pressure 800 bar, for use with multicolumn thermostat and 1290 Infin ity valve drive 1 * 1 items
Catalog # HEWL5067-4284
Supplier:  Agilent
InfinityLab Quick Change valve head, 6-column selector, maximum pressure 800 bar, for use with multicolumn thermostat and 1290 Infin ity valve drive 1 * 1 items
Catalog # HEWL5067-4284
Supplier:  Agilent
Frequently Bought Together