Order Entry
Slovenia
ContactUsLinkComponent
Stericlin® White Line pouches, non-woven polypro / film, very resistant, suitable for STEAM 134°C, EO, FORM and PLASMA, cleanliness level 200 per IEST-STD-CC1246, 43x52cm, DIN EN ISO 11607, case of 5 packs x 100pcs 1 * 500 items
Catalog # 129-1471
Stericlin® White Line pouches, non-woven polypro / film, very resistant, suitable for STEAM 134°C, EO, FORM and PLASMA, cleanliness level 200 per IEST-STD-CC1246, 43x52cm, DIN EN ISO 11607, case of 5 packs x 100pcs 1 * 500 items
Catalog # 129-1471
Frequently Bought Together