Order Entry
Slovenia
ContactUsLinkComponent
Grindomix GM200 Knife Mill GRINDOMIX GM 200 complete with 1 litre grinding container of autoclavable plastic, standard lid and knife of stainless steel 1 * 1 items
Catalog # 412-0329
Supplier:  Retsch GmbH
Grindomix GM200 Knife Mill GRINDOMIX GM 200 complete with 1 litre grinding container of autoclavable plastic, standard lid and knife of stainless steel 1 * 1 items
Catalog # 412-0329
Supplier:  Retsch GmbH
Frequently Bought Together