Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Body lotion, Lindesa®
Body lotion, Lindesa®
Body lotion, Lindesa®, 100 ml tube
Catalog # FAWE13640-008
Supplier:   FAWECO HAUTSCHUTZ
Body lotion, Lindesa®
Catalog # FAWE13640-008
Supplier:  FAWECO HAUTSCHUTZ
Body lotion, Lindesa®
Body lotion, Lindesa®
Body lotion, Lindesa®, 100 ml tube
Price
404,00 €
Log in for availability
1 * 24 items
Product Details & Documents
Barrier cream and care cream with beeswax is designed to care for skin exposed to minor contamination, and is also suitable for after-work care.
  • Lightly greasy, absorbs quickly
  • Special care ingredients improve the skin's elasticity and suppleness
  • Does not affect the sense of touch
  • Emulsion type: O/W, neutral pH level, silicone-free, scented
The cream strengthens the skin's immunity, prevents skin damage from exposure to external contamination, and has a soothing effect on irritated skin. It smooths and nourishes without any long-lasting, greasy film.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Description:
    Body lotion, Lindesa®, 100 ml tube
  • Seq:
    02