Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Adenosine 5-Triphosphate (ATP) - 1 ml 1 * 1 mL
Catalog # NEWEP0756S
Supplier:
NEW ENGLAND BIOLABS
Expand image
Adenosine 5-Triphosphate (ATP) - 1 ml 1 * 1 mL
Catalog # NEWEP0756S
Supplier:
NEW ENGLAND BIOLABS
Frequently Bought Together
MHC Class I Heavy Chain (Restricted expression) Mouse Monoclonal Antibody [Clone ID: HCA2] 1 * 100 µG
Supplier: OriGene
PathHunter eXpress CHRM5 Activated GPCR Internalization Assay 1 * 1 KIT
Supplier: DISCOVERX
InCELL Hunter eXpress CLK2 Kinase Binding Assay 1 * 1 KIT
Supplier: DISCOVERX
PathHunter eXpress MRGPRF CHO-K1 β-Arrestin Orphan GPCR Assay 1 * 1 KIT
Supplier: DISCOVERX
E.Z.N.A.® Mollusc & Insect DNA Kit
Supplier: OMEGA BIO-TEK
PathHunter eXpress BDKRB1 CHO-K1 β-Arrestin GPCR Assay 1 * 1 KIT
Supplier: DISCOVERX
PATHHUNTER U2OS FGFR1 FUNCTIONAL ASSAY 1 * 2 Vial
Supplier: DISCOVERX
Taq Dna Ligase 2,000 units 1 * 2 KU
Supplier: NEW ENGLAND BIOLABS