NEBNext® dsDNA Fragmentase® - 250 assays 1 * 250 AU
Catalog # NEWEM0348L
Supplier: NEW ENGLAND BIOLABS
NEBNext® dsDNA Fragmentase® - 250 assays 1 * 250 AU
Catalog # NEWEM0348L
Supplier: NEW ENGLAND BIOLABS
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Restriction Endonucleases Sap I 1250 units 1 * 1.250 EU
Supplier: NEW ENGLAND BIOLABS
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF