TISSUE MOBILO COMBITEX PU RED 1 * 1 items
Catalog # BSNC19791
Supplier: Basan
Frequently Bought Together
VWR® pHenomenal® pH Electrodes
Supplier: VWR Collection
Lens cleaning tissues
Supplier: HONEYWELL SAFETY
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
VWR®, Bench Protectors
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Kitchen rolls, Tork Premium
Supplier: Essity
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Facial tissues, KLEENEX®
Supplier: KIMBERLY CLARK