LITHIUM HEXAFLUOROPHOSPHATE SOLUTION IN 1 * 100 mL
Catalog # SIAL746789-100ML
Supplier: Merck
Frequently Bought Together
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific