C-mount adapter model U-TV0.63XC-1-8 with magnification factor 0.63x. Compatible with SZX, SZX2, CKX, CX, BX2, BX3, BXiS, MX trinocluar tubes, IX2/IX3 left side port, IX3 right side port, GX intermediate side port (GX-SPU) 1 * 1 items
Catalog # OLYMN5667800
Supplier: OLYMPUS OPTICAL
Frequently Bought Together