Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-17A

Mouse Recombinant IL-17A

25 µG
Catalog # STMC78033.1
Mouse Recombinant IL-17A
Catalog # STMC78033.1
CAS Number:  
Mouse Recombinant IL-17A
Mouse Recombinant IL-17A

25 µG

CAS Number:  
Price
275,00 €
Log in for availability
1 * 25 µG
Product Details & Documents

Interleukin 17A (IL-17A) is the founding member of the family of cytokines that includes Interleukin 17B through Interleukin 17F. It is a potent proinflammatory cytokine that plays a key role in defense against pathogens. IL-17A and IL-17F signal as homodimers or heterodimers through the same receptor, and activate NF-kB, MAPK, and C/EBP pathways (Gaffen). IL-17A receptor is expressed on a variety of cell types, including hematopoietic cell compartments. IL-17A is produced by T helper 17 cells, CD8+ T cells, γδ T cells, natural killer T cells, B cells, neutrophils, innate lymphoid cells and mesenchymal stromal cells (MSCs; Zenobia and amp; Hajishengallis; Mojsilovic et al.). IL-17A receptor is expressed at particularly high levels on stromal cells, including MSCs. IL-17A increases the frequency and the average size of colony-forming units-fibroblast derived from bone marrow, as well as the proliferation of bone marrow-derived MSCs. IL-17A suppresses osteogenic differentiation and bone formation of bone marrow-derived MSCs. The action of IL-17A on hematopoiesis is deeply reliant on the microenvironment and the induction of other regulators. In healthy mouse bone marrow, IL-17A stimulates myeloid and early stage erythroid progenitor cells but inhibits late stage erythroid progenitor cells (Mojsilovic et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    25 µG
  • Cat. No.:
    STMC78033.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-17A
  • Purity:
    ≥95%
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.