Micro Kjeldahl 100 ml flasks for use with the Micro Digestor. They feature long necks, rounded bottoms and thick walls. Borosilicate glass. 1 * 6 items
Micro Kjeldahl 100 ml flasks for use with the Micro Digestor. They feature long necks, rounded bottoms and thick walls. Borosilicate glass. 1 * 6 items