Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Mobile laptop workstation

Mobile laptop workstation

Mobile laptop workstation on casters, 500×500×800 mm
Catalog # PALBWP0504
Supplier:   Palbam
Mobile laptop workstation
Catalog # PALBWP0504
Supplier:  Palbam
Mobile laptop workstation
Mobile laptop workstation

Mobile laptop workstation on casters, 500×500×800 mm

Price
2 760,00 €
Log in for availability
1 * 1 items
Product Details & Documents

The mobile laptop workstation provides a minimum footprint, stable work platform for working with laptops within the cleanroom.

  • Made of 304L stainless steel with electropolished finish
  • Custom fabricated to fit based on laptop dimensions

Weighted base ensures stability and is equipped with cleanroom casters with brakes. The slots on the surface allowing the laptop to be secured to workstation if required.

Packaging: These can be shipped flat-packed and assembled by customer in 10 minutes.

Documents

Specifications
  • Description:
    Mobile laptop workstation on casters
  • Seq:
    1
  • W×D×H:
    500×500×800 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.