Order Entry
Slovenia
ContactUsLinkComponent
Gas-tight option model - incl. additional measures for the greatest possible impermeability to gas not in conjunction with access p ort or interior lighting option 1 * 1 items
Catalog # BIND8012-1688
Supplier:  Binder
Gas-tight option model - incl. additional measures for the greatest possible impermeability to gas not in conjunction with access p ort or interior lighting option 1 * 1 items
Catalog # BIND8012-1688
Supplier:  Binder
Frequently Bought Together