PROCARE LAB 11 MA, 10 KG 1 * 10 kg
Catalog # MIEL10490580
Supplier: MIELE
Frequently Bought Together
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® Disposable Spatulas, PP
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Bromophenol blue 0.04% (w/w) in aqueous solution, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY