Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Disinfectant solutions, Bacillol AF

Disinfectant solutions, Bacillol AF

Rapid Disinfectant Solution, Bacillol AF, 5 litre plastic container
Catalog # BODE9802174
Disinfectant solutions, Bacillol AF
Catalog # BODE9802174
Supplier:  
Disinfectant solutions, Bacillol AF
Disinfectant solutions, Bacillol AF

Rapid Disinfectant Solution, Bacillol AF, 5 litre plastic container

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SURE CLEANER DEGREASER 2x5L W1779 1 * 2 Bottle Catalog #141-1587

Product 141-1587 is similar to BODE9802174 but manufacturer and specifications may vary.

Product Details & Documents

Alcohol based disinfectant, aldehyde-free and fragrance-free, suitable for spray wipe of all surfaces resistant to alcohol in the agro-food and pharmaceutical industries, in hospital environments or in medical practices. Suitable for application where rapid disinfection is required that leaves no residue.

Contents:
Propan-1-ol 450 mg/g
Propan-2-ol 250 mg/g
Ethanol 47 mg/g

Caution: Use biocides safely. Always read the label and product information before use.

Avantor recommend that you read the package insert and the instructions prior to each use of this product.

Documents

Specifications
  • Description:
    Rapid Disinfectant Solution, Bacillol AF
  • Seq:
    08
  • Packed:
    5 litre plastic container
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Universal sprayhead for 500 ml and 1000 ml bottles
    Universal sprayhead for 500 ml and 1000 ml bottles
    BODL8158704
    Log in for availability
    1 * 1 items