V-SOLV ICP SOLVENT 5 GALLON IS A PROPRIETARY SOLVENT THAT IS USED FOR DILUTING OIL AND OTHER ORGANIC LIQUIDS FOR ANALYSIS BY ICP AND ICP-MS. USE V-SOLV AS A MATRIX BLANK AND AS A DILUENT FOR YOUR CALIBRATION STANDARDS AND SAMPLES FOR OUTSTANDING NEB 1 * 1 items
Catalog # PERCN9308378
Supplier: PerkinElmer
Frequently Bought Together