ETALON MERCURE 100MG/L DANS 5% HNO3 Mercury Hg - 100 mg/l in 5% HNO3 for ICP-MS (HgO 99.999+%). Duree de conservation : 12 mois a ouverture du flacon. Duree de validite: 24 mois. 1 * 100 mL
Catalog # CMPLM324.5NP.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together