Order Entry
Slovenia
ContactUsLinkComponent
Anti-TP53 antibody produced in mouse purified immunoglobulin, buffered aqueous solution species reactivity ,,human application(s) ,,indirect immunofluorescence: suitable ,, western blot: suitable clone ,,polyclonal concentration ,,~1 mg/mL antibody f 1 * 50 µG
Catalog # SIALSAB1406544-50U
Supplier:  Merck
Anti-TP53 antibody produced in mouse purified immunoglobulin, buffered aqueous solution species reactivity ,,human application(s) ,,indirect immunofluorescence: suitable ,, western blot: suitable clone ,,polyclonal concentration ,,~1 mg/mL antibody f 1 * 50 µG
Catalog # SIALSAB1406544-50U
Supplier:  Merck
Frequently Bought Together