Multi-parameter meters, handheld, MultiLine® Multi 3620 IDS / 3630 IDS
Product Details & Documents
These compact, digital precision meters enable measurements of pH, ORP, conductivity, dissolved oxygen (DO) and turbidity. Multi 3620 IDS offers two measurement inputs while Multi 3630 IDS offers three. The measuring channels are freely combinable for same or different parameters. Their the field and in the laboratory.robust and waterproof design make these meters ideal for measurements in.
- Simultaneous measurement of two or three parameters
- MultiLab® Importer software enables data acquisition via Excel®
- QSC function for optimum tracking of sensor quality
- CMC function (continuous measurement control) enables you instantly evaluate the current measured value
- Calibration records with history and additional information are stored in IDS sensors
- Automatic sensor recognition (IDS sensors only) enables usage of different IDS sensors on one meter without recalibration
- Meters can be used with IDS sensors (standard or wireless ready) and with analogue sensors (optional IDS adapter required)
- Available as sets in cases with sensors and accessories
Wireless ready: With the aid of the adapters, IDS sensors with plug head connectors (variant P) can be wirelessly connected to these meters. This cable-free operation is very convenient for hard to access locations.
Delivery information: Meters only (without sensors) are supplied with 4 NiMH rechargeable batteries (1,2 V Mignon type AA), USB cable (A plug on mini B plug), power pack, short instructions, detailed operating manual (4 languages), CD-ROM with USB drivers, manual and Software MultiLab® Importer.
The sets also include sensors and/or specific accessories.
Please note: A wireless module for the sensor and for the meter is required for the wireless operation. These modules need to be ordered separately.
Documents
MultiLine® IDS Portable Meters
Product Literature
Multiparameter Profile Measurement with IDS.pdf
Additional Product Details
WTWA2FD570_TDS_EN
Datasheet
Specifications
- Description:Multi-parameter meter (3 measurement inputs), Multi 3630 IDS, set G with SenTix® 940-3 pH electrode, TetraCon® 925-3 conductivity cell, FDO® 925-3 optical DO sensor, case and armoring SM Pro
- Seq:07
- mV range:±2500 / ±1200,0
- Compliance:IP 67
- pH range:−2,000...+20,000
- Data logging capacity:500 (manual), 10000 (automatic)
- Power supply:Charger (100 - 240 V, 50 - 60 Hz), 4×1,2 V NiMH batteries (type AA), or via Mini USB-B
- Temperature compensation:Non linear, linear, off
- DO concentration:0,00 - 20,00 / 0,0 - 90,0 mg/L
- Weight:0,4 kg
- Cat. No.:662-2145
- Turbidity range:0 mg/l - 199,9 g/l
- Input:3
- Data logging - log function:Manual or automatic
- Model:Multi 3630 IDS
- Conductivity range:0,001 µS/cm - 1000 mS/cm
- Outputs:Mini-USB-B, USB-A
- W×D×H:180×80×55 mm
- Display:LCD, colour graphic, backlit
Accessories
Product Information
- Catalog #AvailabilityPrice
- USB charger for wireless modules
Product Information
- Catalog #AvailabilityPrice
- Case, KS MultiLine® 1, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °CCase, KS MultiLine® 1, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C662-2133Log in for availability1 * 1 items - 292,00 €
Product Information
- Catalog #AvailabilityPrice
- Kit consisting of wireless module for sensor and meter (1 of each), USB charger and universal USB power supplyKit consisting of wireless module for sensor and meter (1 of each), USB charger and universal USB power supply662-2050Log in for availability1 * 1 items - 448,00 €
Product Information
- Catalog #AvailabilityPrice
- QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)662-2132Log in for availability1 * 1 items - 38,60 €
Product Information
- Catalog #LengthAvailabilityPrice
- AS/IDS-1,5, connection cable for IDS sensors with waterproof plug headAS/IDS-1,5, connection cable for IDS sensors with waterproof plug head662-17331,5 mLog in for availability1 * 1 items - 114,00 €
Product Information
- Catalog #AvailabilityPrice
- Large field case, KS MultiLine® 2, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °CLarge field case, KS MultiLine® 2, without meter/sensors, incl. armoring SM Pro, buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413 µS/cm at 25 °C662-2134Log in for availability1 * 1 items - 328,00 €
Product Information
- Catalog #AvailabilityPrice
- IDS wireless module for IDS meters, IDS WLM-MIDS wireless module for IDS meters, IDS WLM-M665-0424Log in for availability1 * 1 items - 164,00 €
Product Information
- Catalog #AvailabilityPrice
- IDS wireless module for plug head sensors, IDS WLM-SIDS wireless module for plug head sensors, IDS WLM-S665-0423Log in for availability1 * 1 items - 204,00 €
Product Information
- Catalog #AvailabilityPrice
- Universal USB power supply
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



