Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Erlenmeyer flasks, baffled, with membrane screw cap

Erlenmeyer flasks, baffled, with membrane screw cap

2000 ml, 265 mm, 166 mm, GL 45
Catalog # 214-0430
Supplier:   SciLabware
Erlenmeyer flasks, baffled, with membrane screw cap
Catalog # 214-0430
Supplier:  SciLabware
Erlenmeyer flasks, baffled, with membrane screw cap
Erlenmeyer flasks, baffled, with membrane screw cap

2000 ml, 265 mm, 166 mm, GL 45

Price
642,00 €
Log in for availability
1 * 5 items
Product Details & Documents

PYREX®, borosilicate glass.

  • Four equidistant baffles provide consistent agitation when used with compatible orbital shakers
  • Excellent chemical resistance, high temperature resistance
  • Flasks can be autoclaved with the membrane screw cap fastened

Ideal for aerobic microbiology and cell culture. Complete with blue polypropylene GL 45 screw cap with a gas-permeable PTFE membrane that permits gaseous exchange whilst contents remain sterile.

Specifications
  • Ext.Ø:
    166 mm
  • Thread:
    GL 45
  • Capacity:
    2000 ml
  • Height:
    265 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.