ChemiDoc-It 2 515 Cam/Dkrm Kit515 camera: 4.1 mp (with fixed lens), 16 bit monochrome, digital camera with mount, with binning capab ilities, cooling to -57C from ambient. MOTORISED fixed 25mm, f/0.95 lens, close up diopter, 50x50mm Ethidium Bromid 1 * 1 items
Catalog # UVPA97-0724-02
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together