SAFECAP Capillary tubes with self-sealing plug, heparinized (Ammonium Heparin), 75 mm 1 * 2.000 ST
Catalog # MSPPH75-2000M
Supplier: MARKET SOURCE PART PROCESS
SAFECAP Capillary tubes with self-sealing plug, heparinized (Ammonium Heparin), 75 mm 1 * 2.000 ST
Catalog # MSPPH75-2000M
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Erlenmeyer flasks with baffles, with or without membrane screw caps, DURAN®
Supplier: DWK Life Sciences
Flasks, round bottom, with standard ground joint
Supplier: Lenz Laborglas GmbH & CO.KG
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR® Erlenmeyer Flasks, with Standard Ground Joint
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
VWR®, Dishes, Crystallising
Supplier: VWR Collection
Funnels with short stem, DURAN®
Supplier: DWK Life Sciences
Stretchable silicone lids for laboratory vessels, DURAN®
Supplier: DWK Life Sciences
VWR®, Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Separating Funnels, Conical
Supplier: VWR Collection
Ampoules, standard, WHEATON®
Supplier: DWK Life Sciences
Screw Caps for Bottles
Supplier: RIXIUS
Ampoules, pre-scored, WHEATON®
Supplier: DWK Life Sciences
Specimen tubes
Supplier: MURRAY S
CAPS SPOUT NS 29/32 1 * 1 items
Supplier: NEUBERT VOLUME GLASSWAERE
Grinding balls for MM 200 and MM 400
Supplier: Retsch GmbH