Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
All-in-One Kit 2, 6x250 mL (1 x 4.01, 7.00, 10.00, Pepsin-HCl, Storage, 3mol/l KCl) 1 * 1 KIT
Catalog # METT30095315
Supplier:
Mettler - Toledo
Expand image
All-in-One Kit 2, 6x250 mL (1 x 4.01, 7.00, 10.00, Pepsin-HCl, Storage, 3mol/l KCl) 1 * 1 KIT
Catalog # METT30095315
Supplier:
Mettler - Toledo
Frequently Bought Together
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.9%, HiPerSolv CHROMANORM® Reag. Ph. Eur., ACS, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals