Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ION STANDARD F 10 MG/L 1 * 500 mL
Catalog # METT30090852
Supplier:
Mettler - Toledo
Expand image
ION STANDARD F 10 MG/L 1 * 500 mL
Catalog # METT30090852
Supplier:
Mettler - Toledo
Frequently Bought Together
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH Standard Solutions, IUPAC, Certified
Supplier: Hach