Order Entry
Slovenia
ContactUsLinkComponent
SICCO Shelf Suitable for Maxi Desiccators, Big Star Desiccators and Desiccator wall, stainless steel, including two polyamide rails. Width x height x depth mm 495 x 0,8 x 515. Material of shelf Edelstahl/stainless steel. Bores 36 x Ø 20 mm. Material 1 * 1 items
Catalog # BOHLV1860-03
Supplier:  Bohlender
undefined
SICCO Shelf Suitable for Maxi Desiccators, Big Star Desiccators and Desiccator wall, stainless steel, including two polyamide rails. Width x height x depth mm 495 x 0,8 x 515. Material of shelf Edelstahl/stainless steel. Bores 36 x Ø 20 mm. Material 1 * 1 items
Catalog # BOHLV1860-03
Supplier:  Bohlender
Frequently Bought Together