Order Entry
Slovenia
ContactUsLinkComponent
Manual mass comparator XPE505C - max. capacity: 520g - readibility: 0,01mg - repeatability ABA at low load (test weight): 0,01mg - repeatability ABA at nominal load (test weight): 0,035mg - OIML Class: E1, E2, F1, F2, M1, M2, M3 - measuring time: 5.0 1 * 1 items
Catalog # METB30137978
Supplier:  Mettler - Toledo
Manual mass comparator XPE505C - max. capacity: 520g - readibility: 0,01mg - repeatability ABA at low load (test weight): 0,01mg - repeatability ABA at nominal load (test weight): 0,035mg - OIML Class: E1, E2, F1, F2, M1, M2, M3 - measuring time: 5.0 1 * 1 items
Catalog # METB30137978
Supplier:  Mettler - Toledo
Frequently Bought Together