ROTAVAPOR SYSTEM R-215 W/ V-855 V-710 ************************************** This system consists of the following components that are already included: - Rotavapor R-215 with V-855, structure V P+G 29/32 with water bath B-491, 20-180 degrees of Cels 1 * 1 ST
Catalog # BUCH23111V12153
Supplier: BUCHI
Frequently Bought Together