Order Entry
Slovenia
ContactUsLinkComponent
Rotavapor R-300 B-305, no joint., C, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Cold trap, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300150C011
Supplier:  BUCHI
Rotavapor R-300 B-305, no joint., C, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Cold trap, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300150C011
Supplier:  BUCHI
Frequently Bought Together