Sodium bromide ACS
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7647-15-6
- Formula:NaBr
- MDL Number:MFCD00003475
- Molecular Weight:102.89 g/mol
- Boiling Pt:1390 °C (760 mmHg)
- Melting Pt:755 °C
- Density:3,2 g/cm³ (20 °C)
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Appearance:Colorless to white Powder or solid
- Titration Argentometric:≥99.0 %
- pH:5.0 to 8.8
- Chloride (Cl):≤0.2 %
- Bromate (BrO3):≤0.001 %
- Sulfate (SO4):≤0.002 %
- Barium (Ba):≤0.002 %
- Calcium (Ca):≤0.002 %
- Iron (Fe):≤0.0005 %
- Magnesium (Mg):≤0.001 %
- Potassium (K):≤0.1 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



