Order Entry
Slovenia
ContactUsLinkComponent
BN-13/75, Adapter set for R-12/10 - Set of 12 adapters - made of POM-C - max. temperature 100°C - for vacutainers 2-5mL - dimensions (Ø x length): 13x75mm 1 * 1 items
Catalog # BSANBS-010208-PK
Supplier:  BIOSAN
BN-13/75, Adapter set for R-12/10 - Set of 12 adapters - made of POM-C - max. temperature 100°C - for vacutainers 2-5mL - dimensions (Ø x length): 13x75mm 1 * 1 items
Catalog # BSANBS-010208-PK
Supplier:  BIOSAN
Frequently Bought Together