Order Entry
Slovenia
ContactUsLinkComponent
PLCN20XPH-1-7 objective Plan Achromat C objective with 20x magnification. With working distance of 1.2 mm, numerical aperture of 0.4 and cover correction 0.17 mm. For phase-contrast observations (PH1). 1 * 1 items
Catalog # OLYMN4239700
Supplier:  OLYMPUS OPTICAL
PLCN20XPH-1-7 objective Plan Achromat C objective with 20x magnification. With working distance of 1.2 mm, numerical aperture of 0.4 and cover correction 0.17 mm. For phase-contrast observations (PH1). 1 * 1 items
Catalog # OLYMN4239700
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together