Columns, D3606 application set
Agilent and Not-On-column, 2 column set
REST83606-810901
: Restek
Packed, 2 column set gives complete resolution of benzene from ethanol, no compromising coelutions and for accurate quantification of benzene and toluene.
- Individually tested for method applicability
- ASTM method D3606 is alternative to TCEP columns
The column set is to accommodate both valve injection and/or syringe injection, inlet is identified on both column 1 and column 2.
Column 1:1,8 m length, 3,17 mm OD, 2,0 mm ID, nonpolar Rtx-1, configured with a 50,8 mm inlet void to facilitate on-column injection.
Column 2:4,9 m length, 3,17 mm OD, 2,0 mm ID, proprietary packing material, inlet of column is identified for proper orientation for connection to the valve.
: Conforms to the specifications established in the current ASTM method D3606 for the quantitation of benzene and toluene in spark ignition fuel containing ethanol.
- Description:Agilent and Not-On-column, 2 column set