Order Entry
Slovenia
ContactUsLinkComponent
C-CHIP NEUBAUER IMPROVED 5 x 50 C-Chips for 500 tests Innovative disposable plastic haemocytometer with Neubauer improved grid. The novel C-Chip combines precise cell counting and ease of use. There is no need for cover slips. Each C-Chip contains tw 1 * 250 items
Catalog # NANODHC-N02
Supplier:  NANOENTEK
C-CHIP NEUBAUER IMPROVED 5 x 50 C-Chips for 500 tests Innovative disposable plastic haemocytometer with Neubauer improved grid. The novel C-Chip combines precise cell counting and ease of use. There is no need for cover slips. Each C-Chip contains tw 1 * 250 items
Catalog # NANODHC-N02
Supplier:  NANOENTEK
Frequently Bought Together