CELLOMETER AUTO M10 Automatic Cell Counter Microscope-based automated cell counter with CCD camera. The system determines cell con- centration and cell size in only 20 µl growth medium or buffer without staining. Viability assays after Trypan blue st 1 * 1 items
Catalog # NEXCCMAM10
Supplier: NEXCELOM BIOSCIENCE
Frequently Bought Together