For all Cellometers - Brightfield only Cellometer Demo Beads. For brightfield only. Compatible with all Cellometers. Volume: 4 x 1 ml Diameter: 10 µm For demos only – No resale !!! Shipment: RT Storage: 4°C 1 * 1 items
Catalog # NEXCB10-02-020
Supplier: NEXCELOM BIOSCIENCE
Frequently Bought Together