ROCKER SHAKER MR-12 Powerful Rocking Shaker Mixing frequency: 1 - 99 rpm Digital Timer: 1 min - 99 h 59 min Tilt angle (1 - 50 rpm adjustable): 1 - 9° Tilt angle (51 - 99 rpm fixed): 10° Non-slip platform: 46 x 36 cm Max. Loading: 5 kg Dimensions: 40 1 * 1 items
Catalog # BSANMR-12
Supplier: BIOSAN
Frequently Bought Together