Order Entry
Slovenia
ContactUsLinkComponent
COMBI-THERM HEATING/COOLING THERMOSTATE for 20 x 2.0 ml tubes Temperature range: -10 (at 20 °C RT) - +100 °C Microprozessor controlled time and temperature Digital timer: 1 min - 96 h Illuminated display Warranty: 2 Years 1 * 1 items
Catalog # BSAN010410-UAI
Supplier:  BIOSAN
COMBI-THERM HEATING/COOLING THERMOSTATE for 20 x 2.0 ml tubes Temperature range: -10 (at 20 °C RT) - +100 °C Microprozessor controlled time and temperature Digital timer: 1 min - 96 h Illuminated display Warranty: 2 Years 1 * 1 items
Catalog # BSAN010410-UAI
Supplier:  BIOSAN
Frequently Bought Together