Potassium Permanganate (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1.000 g
Catalog # APPC141527.1211
Supplier: PanReac AppliChem
Potassium Permanganate (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1.000 g
Catalog # APPC141527.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Lead(II) acetate trihydrate 99.5-103.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium hydrogen sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Nickel(II) sulphate hexahydrate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hydrometers for low and high surface tension
Supplier: Alla France
Potassium hexacyanoferrate(III) ≥98%
Supplier: Thermo Fisher Scientific