Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SULPHATE NO.1 TABLETS IN PACK OF 100 1 * 100 items
Catalog # LOVI515220BT
Supplier:
Lovibond
Expand image
SULPHATE NO.1 TABLETS IN PACK OF 100 1 * 100 items
Catalog # LOVI515220BT
Supplier:
Lovibond
Frequently Bought Together
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Test strips, nitrite, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY