Order Entry
Slovenia
ContactUsLinkComponent
MultiRAE Lite Pumped O2/CO/H2S/Nh3/IR-LEL Size: 193 x 96,5 x 66 mm Weight: 880g Operation Temperature: -20 bis 50C Languages: Arabic, English, German, Chinese, Russian, Japanese, and more. Sensors: O2: electrochemical Oxygen-Sensor, Range: 0-30% Vol 1 * 1 items
Catalog # RAESMAA3B40128E010
Supplier:  RAE SYSTEMS
MultiRAE Lite Pumped O2/CO/H2S/Nh3/IR-LEL Size: 193 x 96,5 x 66 mm Weight: 880g Operation Temperature: -20 bis 50C Languages: Arabic, English, German, Chinese, Russian, Japanese, and more. Sensors: O2: electrochemical Oxygen-Sensor, Range: 0-30% Vol 1 * 1 items
Catalog # RAESMAA3B40128E010
Supplier:  RAE SYSTEMS
Frequently Bought Together