Order Entry
Slovenia
ContactUsLinkComponent
qPCR detection of Theileria parva. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endoge nous control, extraction control 1 * 150 Tests
Catalog # PATH-T.PARVA
Supplier:  PRIMERDESIGN
qPCR detection of Theileria parva. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endoge nous control, extraction control 1 * 150 Tests
Catalog # PATH-T.PARVA
Supplier:  PRIMERDESIGN
Frequently Bought Together