qPCR detection of Coronavirus 2012 genomes. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curv e, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HCOV-2012
Supplier: PRIMERDESIGN
Frequently Bought Together