Order Entry
Slovenia
ContactUsLinkComponent
Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm), angled, for exhaust air ducting (if necessary for connection by hose), on ly models U, I, S 1 * 1 items
Catalog # MEMMB29719UF1060PL
Supplier:  MEMMERT
Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm), angled, for exhaust air ducting (if necessary for connection by hose), on ly models U, I, S 1 * 1 items
Catalog # MEMMB29719UF1060PL
Supplier:  MEMMERT
Frequently Bought Together