Order Entry
Slovenia
ContactUsLinkComponent
VZV-gE (aa 48-135) protein.The protein contains the VZV-gE immunodominant regions, amino acids 48-135.The protein is purified by pro prietary chromatographictechnique.Varicella-zoster virus (VZV) is an extremely cell-associatedalpha herpesvi 1 * 1 mg
Catalog # BIRBORB84893-1
Supplier:  Biorbyt
VZV-gE (aa 48-135) protein.The protein contains the VZV-gE immunodominant regions, amino acids 48-135.The protein is purified by pro prietary chromatographictechnique.Varicella-zoster virus (VZV) is an extremely cell-associatedalpha herpesvi 1 * 1 mg
Catalog # BIRBORB84893-1
Supplier:  Biorbyt