Order Entry
Slovenia
ContactUsLinkComponent
TGFB3 Human Recombinant produced in plant is disulfide-linked homodimeric, glycosylated, polypeptide chain containing 118 amino acid s and having a molecular mass of 27.2kDa. The TGFB3 is fused to 6xHis tag at N-terminus and purified by stan 1 * 1 µG
Catalog # BIRBORB80615-1
Supplier:  Biorbyt
TGFB3 Human Recombinant produced in plant is disulfide-linked homodimeric, glycosylated, polypeptide chain containing 118 amino acid s and having a molecular mass of 27.2kDa. The TGFB3 is fused to 6xHis tag at N-terminus and purified by stan 1 * 1 µG
Catalog # BIRBORB80615-1
Supplier:  Biorbyt