Order Entry
Slovenia
ContactUsLinkComponent
Thermostatically controlled incubator model TC 256 G contains 4 metal racks + 1 bottom grid + 7 sockets. Net capacity appr. 255l. Temp. range 2 to 40C in steps of 0.1C. Ideal for BOD determination at 20 C. Model with glass door.inner size : 470 B 1 * 1 items
Catalog # LOVI2438235
Supplier:  Lovibond
undefined
Thermostatically controlled incubator model TC 256 G contains 4 metal racks + 1 bottom grid + 7 sockets. Net capacity appr. 255l. Temp. range 2 to 40C in steps of 0.1C. Ideal for BOD determination at 20 C. Model with glass door.inner size : 470 B 1 * 1 items
Catalog # LOVI2438235
Supplier:  Lovibond
Frequently Bought Together