Thermostatically controlled incubator model TC 135 S contains 3 metal racks + 1 bottom grid + 4 sockets. Net capacity appr. 135l. Temp. range 2 to 40C in steps of 0.1C. Ideal for BOD determination at 20 C. Model with standard door. inner size : 51 1 * 1 items
Catalog # LOVI2438200
Supplier: Lovibond
Frequently Bought Together