Order Entry
Slovenia
ContactUsLinkComponent
CO2 ROLL-IN INCUBATOR, 230VOLTS, CE CORD, 40 CUBIC FT.,FOR ROLLER BOTTLE APPARATUS AND HIGH VOLUME TISSUE CULTURE APPLICATIONS AND CELL HARVESTING 1 * 1 items
Catalog # WHEAWI057607-C-E
Supplier:  DWK Life Sciences
undefined
CO2 ROLL-IN INCUBATOR, 230VOLTS, CE CORD, 40 CUBIC FT.,FOR ROLLER BOTTLE APPARATUS AND HIGH VOLUME TISSUE CULTURE APPLICATIONS AND CELL HARVESTING 1 * 1 items
Catalog # WHEAWI057607-C-E
Supplier:  DWK Life Sciences
Frequently Bought Together