Order Entry
Slovenia
ContactUsLinkComponent
Mp s/chamber, inert single pass cyclonic. 1/pk. inert single pass cyclonic spray chamber used with agilent 41/42 mp-aes. recommended for use with hf digests and high tds samples 1 * 1 items
Catalog # HEWLG8000-70008
Supplier:  Agilent
Mp s/chamber, inert single pass cyclonic. 1/pk. inert single pass cyclonic spray chamber used with agilent 41/42 mp-aes. recommended for use with hf digests and high tds samples 1 * 1 items
Catalog # HEWLG8000-70008
Supplier:  Agilent
Frequently Bought Together