Order Entry
Slovenia
ContactUsLinkComponent
TRAY CHIP 4 INCH Pocket size: 4,4 x 4,4 x 0,4mm STAT PRO400 dark green = -74D05D All trays with bumps on backside of the tray H44-173-25-74D05D 1 * 50 ST
Catalog # ENTEH441732574D05D
Supplier:  ENTEGRIS
undefined
TRAY CHIP 4 INCH Pocket size: 4,4 x 4,4 x 0,4mm STAT PRO400 dark green = -74D05D All trays with bumps on backside of the tray H44-173-25-74D05D 1 * 50 ST
Catalog # ENTEH441732574D05D
Supplier:  ENTEGRIS
Frequently Bought Together